Learn More
American Research Products Inc Mouse Histidine rich glycoprot
Catalog No. 5000327012
Encompass
Mouse Histidine-rich glycoprotein (HRG) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27012 Supplier American Research Products Inc Supplier No. CSBEL010736MO
Mouse Histidine-rich glycoprotein (HRG) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma tissue homogenates Method Competitive