Learn More
Biomatik Corporation Mouse Histidine Rich Glycoprotein (HRG) ELISA Kit
Supplier:  Biomatik Corporation EKU04806
Encompass_Preferred
Mouse Histidine Rich Glycoprotein (HRG) ELISA Kit for detecting HRG in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.44ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ESB3. Gene ID: 94175. Target Synonyms: HPRG; HRGP; Histidine-Proline Rich Glycoprotein; Thrombophilia Due To Elevated HRG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5600