Learn More
American Research Products Inc Mouse Histone deacetylase comp
Catalog No. 5000328197
Encompass
Mouse Histone deacetylase compleX subunit SAP130 (SAP130) ELISA kit from CUSABIO Detection Range 46 88 pg/mL-3000 pg/mL Sensitivity 11 72 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28197 Supplier American Research Products Inc Supplier No. CSBEL020694MO
Mouse Histone deacetylase compleX subunit SAP130 (SAP130) ELISA kit from CUSABIO Detection Range 46 88 pg/mL-3000 pg/mL Sensitivity 11 72 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich