Learn More
Sino Biological Mouse HK2 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 58327M08B50
Encompass_Preferred
A DNA sequence encoding the Mouse HK2 (O08528) (Met1-Arg917) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3086