Learn More
Biomatik Corporation Mouse HLA-B Associated Transcript 3 (BAT3) ELISA Kit
Supplier:  Biomatik Corporation EKU04830
Encompass_Preferred
Mouse HLA-B Associated Transcript 3 (BAT3) ELISA Kit for detecting BAT3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z1R2. Gene ID: 224727. Target Synonyms: BAG6; Protein G3; Protein Scythe; BCL2-associated athanogene 6; BAG family molecular chaperone regulator 6; Large proline-rich protein BAG6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6757