Learn More
Biomatik Corporation Mouse Homeobox Protein B4 (HOXB4) ELISA Kit
Supplier:  Biomatik Corporation EKU12247
Encompass_Preferred
Mouse Homeobox Protein B4 (HOXB4) ELISA Kit for detecting HOXB4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10284. Gene ID: 15412. Target Synonyms: HOX2; HOX-2.6; HOX2F. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7296