Learn More
American Research Products Inc Mouse Hormone sensitiVe lipase
Catalog No. 5000326944
Encompass
Mouse Hormone-sensitiVe lipase (LIPE) ELISA kit from CUSABIO Detection Range 0 156 U/mL-10 U/mL Sensitivity 0 039 U/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26944 Supplier American Research Products Inc Supplier No. CSBEL012975MO
Mouse Hormone-sensitiVe lipase (LIPE) ELISA kit from CUSABIO Detection Range 0 156 U/mL-10 U/mL Sensitivity 0 039 U/mL Sample serum plasma tissue homogenates Method Sandwich