Learn More
Raybiotech Inc Mouse HPGD ELISA Kit, 1 Plate Kit, Quantitative, Colorimetric detection, Sensitivity 1.36 ng/ml, Detection range 1.36 ng/ml - 400 ng/ml, Compatible with Cell Culture, Plasma, Serum
Supplier:  Raybiotech Inc ELMHPGD1
Encompass_Preferred
The RayBio Mouse HPGD ELISA Kit is a quantitative, sandwich-based enzyme-linked immunosorbent assay designed for the detection of mouse 15-hydroxyprostaglandin dehydrogenase in biological samples.
  • Designed to detect HPGD in various sample types
  • Colorimetric method of detection
  • Quantitative measurement capability
  • Sensitivity of 1.36 ng/ml
  • Detection range from 1.36 ng/ml to 400 ng/ml
  • Compatible with cell culture supernatants, plasma, and serum
  • Utilizes a 96-well microplate solid support
  • Stable storage at -20°C
  • Estimated lead time of 1-2 business days
  • Available in 1 Plate Kit, 2 Plate Kit, or 5 Plate Kit sizes
  • Shipped with blue ice to maintain integrity
Catalog No. NC2468640