Learn More
Sino Biological Anti-15-PGDH Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50531RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse HPGD / 15-PGDH (rM HPGD / 15-PGDH; Catalog#50531-M08E; Q8VCC1; Met 1-Ser 269). HPGD / 15-PGDH specific IgG was purified by Mouse HPGD / 15-PGDH affinity chromatography.
Catalog No. 50-240-8124