Learn More
Sino Biological Anti-Hemopexin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50271RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Hemopexin (rM Hemopexin; Catalog#50271-M08H; NP_059067.2; Met 1-Gln 460). Hemopexin specific IgG was purified by mouse Hemopexin affinity chromatography.
Catalog No. 50-240-8714