Learn More
Sino Biological Anti-GRP94/HSP90B1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106461T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse HSP90B1, and purified by antigen affinity chromatography.
Catalog No. 50-240-8924