Learn More
Sino Biological Anti-HSPB7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103814T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse HSPB7, and purified by antigen affinity chromatography.
Catalog No. 50-241-0361