Learn More
Sino Biological Anti-HSP60 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50596T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse HSP60 (rM HSP60; Catalog#50596-M07E; NP_034607.3; Leu2-Phe573). HSP60 specific IgG was purified by Mouse HSP60 affinity chromatography.
Catalog No. 50-255-3192