Learn More
Biomatik Corporation Mouse Hypoxia Inducible Factor 1 Alpha (HIF1a) CLIA Kit
Supplier:  Biomatik Corporation EKU09504
Encompass_Preferred
Mouse Hypoxia Inducible Factor 1 Alpha (HIF1a) CLIA Kit for detecting HIF1a in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.11pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q61221. Gene ID: 15251. Target Synonyms: HIF1-A; MOP1; PASD8; Basic Helix-Loop-Helix Transcription Factor; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; Class E basic helix-loop-helix protein 78. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5395