Learn More
BIOHIPPO Mouse I-PTH Microsample
SDP
Catalog No. 5000315538
Encompass
Mouse I-PTH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of I-PTH in serum plasma and other biological fluids It provides a detection range of 12 5-800 pg/mL with a sensitivity of 3 96 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse I-PTH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse I-PTH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse I-PTH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15538 Supplier BIOHIPPO Supplier No. BHE15211982
Mouse I-PTH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of I-PTH in serum plasma and other biological fluids It provides a detection range of 12 5-800 pg/mL with a sensitivity of 3 96 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse I-PTH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse I-PTH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse I-PTH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color