Learn More
Sino Biological Recombinant Anti-ICAM-1/CD54 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50440R009P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD54 / ICAM1 (rM CD54 / ICAM1; Catalog#50440-M08H; NP_034623.1; Met 1-Asn 485) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-1200