Learn More
Sino Biological Recombinant Anti-ICAM-1/CD54 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50440R009100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ICAM-1 / CD54 (rM ICAM-1 / CD54; Catalog#50440-M08H; NP_034623.1; Met1-Asn485).
Catalog No. 50-241-1549