Learn More
Sino Biological Mouse ICAM1 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 50440R10120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ICAM-1 / CD54 (rM ICAM-1 / CD54; Catalog#50440-M08H; NP_034623.1; Met1-Asn485).
Catalog No. 50-237-1955