Learn More
Sino Biological Anti-ICAM-1/CD54 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50440T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ICAM-1/CD54 (rM ICAM-1/CD54; Catalog#50440-M08H; NP_034623.1; Met1-Asn485). ICAM-1/CD54 specific IgG was purified by Mouse ICAM-1/CD54 affinity chromatography.
Catalog No. 50-240-9344