Learn More
Sino Biological Mouse ICAM1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 50440M49HB100
Encompass_Preferred
A DNA sequence encoding the Mouse ICAM1 (P13597) (Met1-Asn485) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-3561