Learn More
Sino Biological Recombinant Mouse ICOS Protein (ECD, hFc & AVI Tag), Biotinylated
Supplier:  Sino Biological 50466M41HB20
Encompass_Preferred
A DNA sequence encoding the Mouse ICOS (NP_059508.2) (Met1-Leu142) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-3269