Learn More
Sino Biological Anti-B7-H2/ICOSLG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50190RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse B7H2 extracellular domain (rM B7H2; Catalog#50190-M08H; NP_056605.1; Met 1-Lys 279). B7H2 specific IgG was purified by mouse B7H2 affinity chromatography.
Catalog No. 50-240-9181