Learn More
PBL Assay Science Mouse Interferon Alpha 1 (IFN-Alpha 1) [1e5 (100,000) units/vial]
Supplier:  PBL Assay Science 121051
Encompass_Preferred
Recombinant protein Mouse Interferon Alpha 1 is manufactured by PBL Assay Science (Catalog #12105-1) for research use only (RUO) and packaged in 1e5 (100,000) units/vial. Mu IFN-Alpha 1 is expressed in E.coli. This cytokine has antiviral, antiproliferative and immunomodulatory activities.
Catalog No. 50-153-7978