Learn More
ACROBiosystems Mouse IFN-alpha 1 Protein, His Tag
Catalog No. 502055274
Encompass_Preferred
Recombinant Protein;1MG;Mouse IFN-alpha 1, His Tag (IFA-M52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Lys 189 (Accession # P01572-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.0 kDa. The protein migrates as 24-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5274 Supplier ACROBiosystems Supplier No. IFAM52H31MG
Recombinant Protein;1MG;Mouse IFN-alpha 1, His Tag (IFA-M52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Lys 189 (Accession # P01572-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.0 kDa. The protein migrates as 24-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.