Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IFN-alpha 2 (Carrier-Free) Recombinant Protein

Catalog No. 509199
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-919-9 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-919-9 Supplier Invitrogen™ Supplier No. 34831285
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Description: IFN alpha2 is a type I interferon, previously known as B-cell interferon, leukocyte interferon, lymphoblast interferon, and pH2-stable interferon. IFN alpha2 is one of at least 23 different known variants of IFN alpha. The individual proteins have molecular masses between 19-26 kDa and consist of proteins with lengths of 156-166 and 172 amino acids. IFN alpha forms are produced by monocytes/macrophages, lymphoblastoid cells, fibroblasts, and a number of different cell types following induction by viruses, nucleic acids, glucocorticoid hormones, and low-molecular weight substances (n-butyrate, 5-bromodeoxyuridine). IFN alpha2 demonstrates antiviral, antiparasitic, antiproliferative activities. Applications Reported: The recombinant mouse IFNa2 has been reported useful for bioassay and ELISA. Applications Tested: The recombinant mouse IFN alpha2 has been tested for inhibition of the cytopathic effect of EMC virus on L929 cells. The ED50 for this effect is typically 0.1 ng/mL, corresponding to a specific activity of 1 x 10e7 Units/mg. Source: E. coli expressed amino acids Cys24-Glu190 of mature mouse IFN alpha2 accession # NM_010503. Bioactivity: The recombinant mouse IFN alpha2 has been tested for inhibition of the cytopathic effect of EMC virus on L929 cells. The ED50 for this effect is typically 0.1 ng/mL, corresponding to a specific activity of 1 x 10e7 Units/mg. Endotoxin: Less than 0.01 ng/ug cytokine as determined by the LAL assay.

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01573
Concentration 0.5 mg/mL
For Use With (Application) Bioactivity, ELISA, Functional Assay
Formulation 50mM sodium acetate with 0.1M NaCl and no preservative; pH 5.0
Gene ID (Entrez) 15965
Molecular Weight (g/mol) 19.4 kDa
Name Mouse IFN-alpha 2 (Carrier-Free)
pH Range 5.0
Purification Method Purified
Quantity 500 μg
Source E. coli
Storage Requirements -80°C
Regulatory Status RUO
Endotoxin Concentration <0.01 ng/μg
Gene Alias alpha-2a interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2
Common Name Interferon alpha-2
Gene Symbol IFNA2
Biological Activity ED50 ̃ 0.1 ng/mL; determined by the inhibition of the cytopathic effect of the EMC virus on L929 cells. This corresponds to a specific activity of 1 x 10e7 Units/mg.
Product Type Protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Sequence Mouse IFN-alpha 2, amino acids Cys24-Glu190 (Accession # NM_010503)
Content And Storage -80°C
Activity ED50 ~ 0.1 ng/mL; determined by the inhibition of the cytopathic effect of the EMC virus on L929 cells. This corresponds to a specific activity of 1x10e7 Units/mg.
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >98% by SDS-PAGE
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.