Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IFN-alpha 2 Recombinant Protein

Catalog No. PI121009
Change view
Click to view available options
Quantity:
>1 x 10^5 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI121009 >1 x 10^5 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI121009 Supplier Invitrogen™ Supplier No. 121009
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

Store at -70°C or below for retention of full activity. Thaw product vial by incubation in cold tap water until just thawed. Aliquot and refreeze unused contents to minimize effects of freeze thawing. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of product should be in buffers containing protein such as 0.1% BSA or tissue culture media with serum. Dilution for freezing of material below 2x10^5 units/mL is not recommended. One freeze thaw cycle is equivalent to thawing an aliquot prepared from the material received.

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01573
Concentration Lot-specific
For Use With (Application) ELISA, Immunoprecipitation, Western Blot
Formulation PBS with 0.1% BSA and no preservative
Gene ID (Entrez) 15965
Molecular Weight (g/mol) 19.3 kDa
Name Mouse IFN-alpha 2
Quantity >1 x 10^5 units
Source E. Coli
Regulatory Status RUO
Gene Alias alpha-2 A interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1 b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 1 b; interferon alpha 2; interferon alpha 2 A; interferon alpha 2 b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2
Common Name Interferon alpha-2
Gene Symbol Ifna1
Biological Activity EC50 ̃ 5 U/mL; determined by use of the cytopathic effect inhibition assay described in [Rubinstein, S. et al. (1981). J. Virol. 37, 755-758 ]; [Familletti, P.C. et al. (1981). Methods in Enzymology, Vol. 78, 387-394 ] with the exception that EMCV rather than VSV was used as the challenge. The activity was determined relative to a lab standard of Mu IFN-alpha-A which was calibrated to the NIH mouse IFN-alpha- standard (Ga02-901-511).
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.