Learn More
PBL Assay Science Mouse Interferon Alpha A (IFN-Alpha A) [1e5 (100,000) units/vial]
Catalog No. 501537977 Shop All PBL Assay Science Products
Encompass_Preferred
Recombinant protein Mouse Interferon Alpha A is manufactured by PBL Assay Science (Catalog #12100-1) for research use only (RUO) and packaged in 1e5 (100,000) units/vial. Mu IFN-Alpha A exhibits high activity and is expressed in E.coli.
Catalog No. 50-153-7977 Supplier PBL Assay Science Supplier No. 121001
Add to Cart
Add to Cart
Recombinant protein Mouse Interferon Alpha A is manufactured by PBL Assay Science (Catalog #12100-1) for research use only (RUO) and packaged in 1e5 (100,000) units/vial. Mu IFN-Alpha A exhibits high activity and is expressed in E.coli.