Learn More
PBL Assay Science VeriKine Mouse Interferon Alpha (IFN-Alpha, IFNa) ELISA, Tissue Culture Media (TCM) [5x96 well plate]
Supplier:  PBL Assay Science 421202
Encompass_Preferred
VeriKine Mouse Interferon Alpha ELISA is manufactured by PBL Assay Science (Catalog #42120-2) for research use only (RUO) and packaged as a 5x96-well plate. This assay detects all 14 Mouse IFN-Alpha subtypes in TCM and has an assay range of 12.5-400 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.
Catalog No. 50-153-8069