Learn More
PBL Assay Science VeriKine Mouse Interferon Beta (IFN-Beta, IFNb) ELISA, Tissue Culture Media (TCM) [1x96 well plate]
Supplier:  PBL Assay Science 424001
Encompass_Preferred
VeriKine Mouse Interferon Beta ELISA is manufactured by PBL Assay Science (Catalog #42400-1) for research use only (RUO) and packaged as a 1x96-well plate. This assay is serum and tissue culture media (TCM) compatible and has an assay range of 15.6-1,000 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 8%.
Catalog No. 50-153-8070