Learn More
Sino Biological Mouse IFN-γ ELISpot Kit (ALP) (Strips) 1 Kit (96 Tests)
Catalog No. 5000696230
Encompass_Preferred
The kit applies for the detection of mouse IFN-γ at the cellular level by counting IFN-γ-secreting cells.
Catalog No. 50-006-96230 Supplier Sino Biological Supplier No. EK50709B1
The kit applies for the detection of mouse IFN-γ at the cellular level by counting IFN-γ-secreting cells.