Learn More
Sino Biological Mouse IFN gamma Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785364
Encompass_Preferred
The Mouse IFN gamma ELISA Pair Set is for the quantitative determination of Mouse IFN gamma.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse IFN gamma coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse IFN gamma present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse IFN gamma monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse IFN gamma present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85364 Supplier Sino Biological Supplier No. SEK507095
The Mouse IFN gamma ELISA Pair Set is for the quantitative determination of Mouse IFN gamma.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse IFN gamma coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse IFN gamma present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse IFN gamma monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse IFN gamma present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.