Learn More
PBL Assay Science DIY (Do-It-Yourself) Mouse Interferon Lambda 2/3 (IFNL2/IFNL3) / Interleukin-28A/B (IL-28A/IL-28B) ELISA, Tissue Culture Media (TCM) [Materials for 15x96 well plate]
Supplier:  PBL Assay Science 628301
Encompass
The Do-It-Yourself (DIY) immunoassay Mouse Interferon Lambda 2/3 (Interleukin-28A/28B) (IL-28A/28B) ELISA kit from PBL Assay Science (Catalog #62830-1) for research use only (RUO) and packaged with materials for 15x96-well plates. This ELISA is Tissue Culture Medium (TCM) compatible and has an assay range of 31.25-2000 pg/ml. This kit contains the basic components necessary for the development of sandwich ELISAs to measure natural and recombinant Mouse IFN-Lambda 2/3 (IL28A/IL28B, respectively). This kit is most reactive with Lambda 2 and Lambda 3.
Catalog No. NC2008879