Learn More
Mabtech Inc Basic ELISpot kit for enumeration of cells secreting IFN-γ
Supplier:  Mabtech Inc 33212A
Encompass
The mouse IFN-γ ELISpot kit is ideal for users who want the flexibility of setting up their own assay. The kit enables enumeration of cells secreting mouse IFN-γ, and contains a matched pair of monoclonal capture and detection antibodies and Streptavidin-ALP.
Catalog No. NC1631304