Learn More
Mabtech Inc Mouse IFN-gamma ELISpot PLUS kit (ALP), 2 plates
Supplier:  Mabtech Inc 33214APW2
Encompass
This kit is ideal for users who want a convenient and sensitive assay. The assay is designed for the enumeration of cells secreting mouse IFN-gamma. The kit includes ELISpot plates pre-coated with monoclonal antibody, biotinylated detection antibody, Streptavidin-ALP and ready-to-use BCIP/NBT-plus substrate.
Catalog No. NC1518531