Learn More
Sino Biological Recombinant Mouse Interferon alpha 1/IFNA1 Protein (His Tag)
Supplier:  Sino Biological 50524M08Y1
Encompass_Preferred
A DNA sequence encoding the mouse Ifna1 (NP_034632.2) (Cys24-Lys189) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3223