Learn More
Sino Biological Anti-Interferon alpha 2/IFNA2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50525T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IFNA2 (rM IFNA2; Catalog#50525-MNAY; NP_034633.2; Cys24-Glu190). IFNA2 specific IgG was purified by Mouse IFNA2 affinity chromatography.
Catalog No. 50-241-0007