Learn More
Sino Biological Recombinant Mouse Interferon alpha 4/IFNA4 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 50672M01H1100
Encompass_Preferred
A DNA sequence encoding the Mouse IFNA4 (NP_034634.1) (Cys25-Glu186) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-255-3156