Learn More
Sino Biological Recombinant Mouse Interferon alpha 4/IFNA4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50672M08H1100
Encompass_Preferred
A DNA sequence encoding the Mouse IFNA4 (NP_034634.1) (Met1-Glu186) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3155