Learn More
Sino Biological Mouse IFNAR1 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 50469R11020
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFNAR1 (rM IFNAR1; Catalog#50469-M08H; NP_034638.2; Met1-Thr429).
Catalog No. 50-237-3571