Learn More
Sino Biological Recombinant Anti-IFN gamma Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50709R348P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFNG / Interferon Gamma (rM IFNG / Interferon Gamma; Catalog#50709-MNAH; NP_032363.1; Met1-Cys155) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-2397