Learn More
Sino Biological Recombinant Anti-IFN gamma Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50709R437100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFN gamma (rM IFN gamma; Catalog#50709-M08H; P01580; Met1-Cys155).
Catalog No. 50-255-3131