Learn More
Sino Biological Mouse IFNG MAb 25TS (Sino Biological)
Supplier:  Sino Biological 40588V07E40100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFN gamma (rM IFN gamma; Catalog#50709-MNAH; NP_032363.1; Met1-Cys155) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-4318