Learn More
Sino Biological Anti-IL-28B/Interferon lambda-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51306T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-28B / IFN-lambda-3 (Catalog#51306-M08H; NP_796370.1; Met1-Val193). IL-28B / IFN-lambda-3 specific IgG was purified by Mouse IL-28B / IFN-lambda-3 affinity chromatography.
Catalog No. 50-241-0441