Learn More
Raybiotech Inc Quantibody Mouse Immunoglobulin Isotyping Array Kit.
Supplier: Raybiotech Inc QAMISO12
Quantitative measurement of 7 Mouse Immunoglubins. Suitable for serum, plasma, or cell culture media samples. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IgA, IgE, IgM, IgG1, IgG2a, IgG2b, IgG3, IgD, two light chains (Kappa and Gamma)