Learn More
Raybiotech Inc Mouse IgE ELISA Kit
Supplier:  Raybiotech Inc ELMIGE1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.14 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects mouse IgE. Other species not determined.
Catalog No. 50-195-0017