Learn More
Crystal Chem Inc Mouse IgE ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80643
Encompass
The Mouse IgE ELISA is used to quantify levels of mouse IgE in serum and plasma using only a 5 µL sample volume. Highlights: Kit uses sample size of only 5 µL. High sensitivity of 1.92 ng/mL. Dynamic range of 6.25 - 400 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7774