Learn More
Crystal Chem Inc Mouse IGF-1 ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80574
Encompass
The Mouse IGF-1 ELISA is used to quantify levels of mouse IGF-1 in plasma, serum, and cell culture using only a 5 µL sample volume. Highlights: Kits use only 5 µL sample, ideal for mouse applications. Dynamic range of 0.5 - 18 ng/mL. High sensitivity of 0.029 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7741