Learn More
FUJIFILM BIOSCIENCES INC Mouse IGF-I (IGF-1), Recombinant Protein, Shenandoah Biotechnology, 50 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 2000550UG
Encompass_Preferred
Insulin-like growth factor I (IGF-I) is a growth factor that is produced by the liver. IGF-1 production is stimulated by growth hormone (GH). IGF-I binds the insulin-like growth factor 1 receptor (IGF1R) and the insulin receptor to stimulate systemic body growth. IGF-I is one of the most potent activators of the AKT signaling pathway, which stimulates cell proliferation and inhibits programmed cell death.
Catalog No. 50-990-805
Edge