Learn More
Raybiotech Inc Mouse IGF-2 ELISA Kit
Supplier:  Raybiotech Inc ELMIGF21
Encompass_Preferred
For Cell Culture Supernatants samples. Sensitivity: 1500 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes mouse IGF-II (for Cell Culture).
Catalog No. 50-195-0019