Learn More
Sino Biological Recombinant Mouse IGF1 Protein
Supplier:  Sino Biological 50437MNAY1
Encompass_Preferred
A DNA sequence encoding the mouse IGF1 (NP_001104744.1) (Gly49-Ala118) was expressed.
Catalog No. 50-255-3273